Imprimer
Scratcher for sampling
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5347-0010 5347-0005 5347-0025
BURK5347-0010EA 1690 EUR
BURK5347-0010 BURK5347-0005 BURK5347-0025
Scratcher for sampling
Samplers Solid Samplers
The scratcher has particularly been designed for sampling cohesive or caked substances.
  • Electro-polished stainless steel
For non pourable substances.