Imprimer
Sample vials, E-C, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
224722 224724 224706 224711 224712 224714 224728 224721 224750 224708 224701 224705 224710 224742 224720 224725 224726 224730 224731 224732 224745 224748 224752 224734 W224810 W226060
215-3572EA 140 EUR
215-3572 215-3573 218-2261 218-2266 218-2267 218-2268 215-3576 215-3571 WHEA224750 WHEA224708 WHEA224701 WHEA224705 WHEA224710 215-3577 WHEA224720 215-3574 215-3575 WHEA224730 WHEA224731 WHEA224732 215-3579 215-3581 WHEA224752 WHEA224734 WHEAW224810 WHEAW226060
Sample vials, E-C, WHEATON®
Fioles
Clear vials made from low extractable borosilicate glass that provides superior chemical resistance. Conform to USP Type I and ASTM E 438 Type I, Class A requirements.
Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products.
Informations de commande: Caps and replacement caps can be purchased separately.
Cdt: Vials packed in two shrink-wrapped packs of 100 vials which reduces contamination from corrugate. The 60 ml vials are packed in one shrink-wrapped pack of 143 vials.