Imprimer
Kits d'instruments de microscopie
Fournisseur: RSG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
066.805 066.807
RSGA066.805EA 117 EUR
RSGA066.805 RSGA066.807
Kits d'instruments de microscopie
Trousses à dissection
Microscopy instrument set in a grey wooden box, all articles are completely made of stainless steel.
  • 1 pair of microscopy scissors
  • 1 microscopy needle straight
  • 1 microscopy needle bent
  • 1 pair of tweezers, pointed straight with guide pin
  • 1 scalpel
Informations de commande: On demand the arrangement of the 7 pcs. set can be individualised.