Imprimer
Flat optical windows
Fournisseur: HELLMA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
665-000-40 201-1-23 202-40 665-000-46 202-46
HELL665-000-40EA 82.8 EUR
HELL665-000-40 HELL201-1-23 HELL202-40 HELL665-000-46 HELL202-46
Flat optical windows
Fenêtres optiques
Optical glass cells and elements dedicated to special applications with detachable window.
Suitable for high precision analytical techniques.