Imprimer
Scintillation vials
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S220 S220-1
720-0496EA 270 EUR
720-0496 548-0256
Scintillation vials
Fioles
These scintillation vials are made of PP. Caps are made of HDPE.
  • Easy removal of caps (¼ turn-twist), leakproof
  • High degree of resistance to organic solvents
  • Shoulderless vial features a full width opening of 23 mm for ease of access
  • Sufficient translucence so that reagent levels or the presence of filter paper can be easily detected
The quality of the sealing system is such that it is not necessary to tighten the closure with a lot of pressure to achieve a leakproof seal. The HDPE closure may not be autoclaved.