Imprimer
Media bottles, borosilicate glass, non-graduated, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
219575 219885 219415 219417 219419 219495 219497 219499 219577 219579 W219580 W219500
WHEA219575EA 318 EUR
WHEA219575 WHEA219885 WHEA219415 WHEA219417 WHEA219419 WHEA219495 WHEA219497 WHEA219499 WHEA219577 WHEA219579 WHEAW219580 WHEAW219500
Media bottles, borosilicate glass, non-graduated, WHEATON®
Flacons Flacons pour milieux de culture
Borosilicate glass, non-graduated.
  • Hand grips provided in 1 L bottle for easy handling
  • Very good chemical resistance
  • Very good temperature resistance
Media bottles are pre-attached with phenolic screw caps with the choice of liner. Without screw caps also available and 125 ml bottles available in amber glass.