Imprimer
Watch dishes
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
643 644 645 646 647
KART643EA 18.4 EUR
KART643 KART644 KART645 KART646 KART647
Watch dishes
Watch Glasses
Made of PTFE, white.
  • Very good chemical, heat and mechanical resistance
  • Heat resistant up to +120 °C
  • For use with foodstuff
For blends or for covering vessels.