Imprimer
Filtre-seringues, Minisart® HY
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16599-HYQ 16596-HYQ 16596-HYK
514-0676EA 1110 EUR
514-0676 512-3222 611-0696
Filtre-seringues, Minisart® HY
Filtres Filtres-seringue
Minisart® HY with chemically inert hydrophobic PTFE is suitable for venting as well as filtration of gases and oils with a pH of 3 - 12. With a pore size of 0,2 µm that can be used for sterile filtration. It has a male luer lock outlet and version HYK is individually packed and sterilised by ethylene oxide (EO). Only available in the EU/EEA and registered countries.
  • Sterile and ready-to-use
  • Minimum extractables or leachables
  • Very low non-specific binding
  • Suitable as venting filter
Individually packed and sterile Minisart® HY syringe filters are optimal for immediate usage whenever sterile filtration or venting is needed.
The low-binding PTFE membrane is the best option for leachable free filtration. Due to its hydrophobic nature the PTFE membrane functions as a water barrier and does not become wetted by aerosols. This keeps the hydrophobic venting filter permeable for gas or air.
The housing of the filter is made of MBS (Methacrylate butadiene styrene) which has a limited compatibility with alcohols but is free of phthalates and passes the plastic class VI test. Housing and membrane are tightly connected which allows bidirectional use.