Imprimer
Biopsy cassettes in QuickLoad™ stacks, Micromesh®, M507T
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M507-8T M507-9T M507-4T M507-10T M507-11T M507-12T M507-2T M507-3T M507-5T M507-6T M507-7T
720-1419EA 826 EUR
720-1419 720-1420 720-1689 720-1411 720-1412 720-1413 720-1414 720-1415 720-1416 720-1417 720-1418
Biopsy cassettes in QuickLoad™ stacks, Micromesh®, M507T
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer.
  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Cdt: Cassettes are packed in stacks of 40. Each case contains 50 stacks for a total of 2000 cassettes.