Imprimer
Flacons, col large, ronds, avec bouchon à vis, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
209627 209628 209629 209630 209665 209666 209667 209668 209625 209550 209669 209425 209626 209427 209428 209545 209546 209547 209548 209549
215-4874EA 224 EUR
215-4874 215-4875 215-4876 215-4877 215-4878 215-4879 215-4880 WHEA209668 215-4872 215-4871 WHEA209669 215-4863 215-4873 215-4864 215-4865 215-4866 215-4867 215-4868 215-4869 215-4870
Flacons, col large, ronds, avec bouchon à vis, WHEATON®
Flacons Flacons en plastique
Ideal for a wide range of general laboratory applications, including packaging, storage, and transportation of liquids. Wide neck bottles are available in LDPE, HDPE, or PP formats.
  • LDPE bottles are flexible, impact-resistant, and chemical-resistant
  • HDPE bottles offer excellent chemical resistance and suitability for freezing at –100 °C
  • High-quality PP bottles offer exellent chemical and temperature resistance
  • Superior linerless double seal closure ensures leak free performance
  • Non-drip pour lip aids safe and clean pouring
  • Moulded tamper proofing bead for use with shrink band
Available as lab packs with polypropylene screw cap assembled in a single twist tied bag and as bulk packed product, double bagged and twist-tied for reduced particulate, with matching closures and bottles bagged separately.
PP bottles are autoclavable, however cap must be completely removed prior to autoclaving.
Bottles can be used as a primary receptacle for the Transport of Diagnostic Specimens as outlined in the IATA Dangerous Goods Regulations, Part 6.3.5. Bottles meet the requirements of packaging for which retention of liquid is a basic function and capable of withstanding the pressure requirements without leakage per DOT CFR 173.27(c), ICAO 4:1.1.6, IATA 5.0.2.9 *Seal psi exceeds 14 psi/1 atmosphere/95 kPA pressure tested. Leak testing results are based on water. Results may vary when packaging other liquids. Customers are advised to test leak resistance to specific storage and transport conditions.