Imprimer
Test tubes with screw caps
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
88202 88205 88200 88201 88210 88212 88207
KART88202EA 431 EUR
KART88202 KART88205 KART88200 KART88201 KART88210 KART88212 KART88207
Test tubes with screw caps
Tubes Tubes à essais
PS, clear, wtih screw cap.
  • Tight screw cap
  • Good chemical resistance
  • Good temperature resistance
Taking into consideration the different applications, they are available sterile single packed, sterile nultiple packed and not sterile.