Imprimer
Disques filtrants, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
251540306 251590208 251510003 251510106 251510209 251510303 251510406 251520004 251520107 251520201 251520304 251520407 251525002 251525105 251525208 251525405 251530005 251530108 251530305 251530408 251540006 251540109 251540203 251540409 251550007 251550101 251550204 251550307 251550401 251560008 251560102 251560205 251560308 251560402 251570009 251570103 251570206 251570309 251570403 251580104 251590002 251590105 251590302 251590405 251620002 251620105 251620208 251620302 251620405 251515207 251525302 251530202 251525508 251515404 251620508 251515507 251555305 251555408 251555005 251555108 251555202 251540503 251550504
SCOT251540306EA 111 EUR
SCOT251540306 SCOT251590208 SCOT251510003 SCOT251510106 SCOT251510209 SCOT251510303 SCOT251510406 SCOT251520004 SCOT251520107 SCOT251520201 SCOT251520304 SCOT251520407 SCOT251525002 SCOT251525105 SCOT251525208 SCOT251525405 SCOT251530005 SCOT251530108 SCOT251530305 SCOT251530408 SCOT251540006 SCOT251540109 SCOT251540203 SCOT251540409 SCOT251550007 SCOT251550101 SCOT251550204 SCOT251550307 SCOT251550401 SCOT251560008 SCOT251560102 SCOT251560205 SCOT251560308 SCOT251560402 SCOT251570009 SCOT251570103 SCOT251570206 SCOT251570309 SCOT251570403 SCOT251580104 SCOT251590002 SCOT251590105 SCOT251590302 SCOT251590405 SCOT251620002 SCOT251620105 SCOT251620208 SCOT251620302 SCOT251620405 SCOT251515207 SCOT251525302 SCOT251530202 SCOT251525508 SCOT251515404 SCOT251620508 SCOT251515507 SCOT251555305 SCOT251555408 SCOT251555005 SCOT251555108 SCOT251555202 SCOT251540503 SCOT251550504
Disques filtrants, DURAN®
Filtres Disques filtrants
Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design
Made from clear DURAN® borosilicate glass 3,3.