Imprimer
Entonnoirs, à usage unique
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F490-3 F490-4 F490-1 F490-2
SIMPF490-3EA 45.1 EUR
SIMPF490-3 SIMPF490-4 SIMPF490-1 SIMPF490-2
Entonnoirs, à usage unique
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
These disposable funnels are made of either polystyrene or polypropylene. Both are tapered at a 60° angle with inside fluting.
  • Good chemical resistance
  • Good temperature resistance
  • PP funnels are autoclavable