Imprimer
Weighing dishes, anti-static
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D250-3 D250-1 D250-2
SIMPD250-3EA 124 EUR
SIMPD250-3 SIMPD250-1 SIMPD250-2
Weighing dishes, anti-static
Boats Weighing Boats
These weighing dishes are safe, contaminant-free, biologically inert economical containers for weighing liquid or powdered samples in the laboratory.
  • Resistant to diluted acids, aqueous solutions, alcohols and bases
  • Flat bottom ensures perfect stability on countertops
  • Smooth surface providing accurate pour-outs with minimal sample loss
  • Facilitate weighing of static-affected samples
  • Ideal for many applications such as weighing, dispensing or storing
Dishes can also be used as quick freeze trays for sample material, discard trays for broken ampules, or mixing trays for small batches. Withstand temperatures up to 80 °C.