Imprimer
Double spatulas, micro, one conical end
Fournisseur: RSG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
039.210 039.160 039.140
RSGA039.210EA 34.4 EUR
RSGA039.210 RSGA039.160 RSGA039.140
Double spatulas, micro, one conical end
Spatulas
Micro double spatulas with one conical end, 7 mm wide. Made of stainless steel.