Imprimer
DURAN® Adapters, Receiver, Vacuum, Straight
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
241312106 241313402 241314604
SCOT241312106EA 79.4 EUR
SCOT241312106 SCOT241313402 SCOT241314604
DURAN® Adapters, Receiver, Vacuum, Straight
Composants de distillation Adaptateurs de distillation
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Straight, with two standard ground joints and screw-on plastic hose connection.