Imprimer
Adapters, receiver, vacuum, bent, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
241302105 241303401 241304603
SCOT241302105EA 85.2 EUR
SCOT241302105 SCOT241303401 SCOT241304603
Adapters, receiver, vacuum, bent, DURAN®
Composants de distillation Adaptateurs de distillation
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent, with two standard ground joints and screw-on plastic hose connection.