Imprimer
Bottles, narrow neck, square, HDPE, with screw caps, LaboPlast®
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0332-0100 0332-1000 0332-0250 0332-0500
BURK0332-0100EA 1.1 EUR
BURK0332-0100 BURK0332-1000 BURK0332-0250 BURK0332-0500
Bottles, narrow neck, square, HDPE, with screw caps, LaboPlast®
Flacons Flacons en plastique
HDPE, transparent.
  • Shatterproof
  • Leakproof
  • Good chemical and temperature resistance
  • Special labelling area for labels or manual lettering
  • Suitable as a disposable bottle