Imprimer
Distributors for distilling receivers, Bredt
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05285204 05285202
271-2542EA 140 EUR
271-2542 271-2541
Distributors for distilling receivers, Bredt
Composants de distillation Receveurs de distillation
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distributors with four cones NS 14/23, for round bottom flasks up to 250 ml volume.