Imprimer
Adapters, receiver, bent, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
243100602 243100808
SCOT243100602EA 31.8 EUR
SCOT243100602 SCOT243100808
Adapters, receiver, bent, DURAN®
Composants de distillation Adaptateurs de distillation
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent, with ground socket.