Imprimer
Microplates, 96-well
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2620 2621 2622 2620/S 2621/S 2622/S
KART2620EA 98.7 EUR
KART2620 KART2621 KART2622 KART2620/S KART2621/S KART2622/S
Microplates, 96-well
Microplaques Microplaques HTS
Multiple-well plates (96 sample wells) in clear PS. Available in three shapes: flat bottom, U bottom and V bottom wells.
  • Aseptic or sterile
  • Single-wrapped
Informations de commande: PS covers are sold separately.