Imprimer
Dropping bottles, LDPE, LaboPlast®
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0306-0030 0306-0100 0306-0050 0306-0500 0306-0010 0306-1000 0306-0250
BURK0306-0030EA 1.1 EUR
BURK0306-0030 BURK0306-0100 215-4290 BURK0306-0500 BURK0306-0010 BURK0306-1000 BURK0306-0250
Dropping bottles, LDPE, LaboPlast®
Flacons Flacons compte-gouttes
LDPE, translucent.
  • Cut off dropper tip before first use
  • Can be reclosed using the captive cap provided
  • Captive cap and dropper cap PE
Dropping bottles are particularly suitable for liquids which must be dispensed in small amounts, drop by drop or as a fine jet.