Imprimer
L-shaped spreaders, PS
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2647 2648
KART2647EA 1310 EUR
KART2647 KART2648
L-shaped spreaders, PS
Étaleurs Étaleurs de cellules
Disposable L-shaped sterile PS spreaders for bacteriology with special 100° angling for easier use. Especially handy for dispersing liquids across agar culture plates.
Cdt: Available single-wrapped or in packs of 5.