Imprimer
Burettes, type Pellet, sans robinet intermédiaire, Schellbach
Fournisseur: HIRSCHMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
344016060 344017060 344017560 344036060 344037060 344037560
612-9831EA 325 EUR
612-9831 HIRS344017060 HIRS344017560 HIRS344036060 HIRS344037060 HIRS344037560
Burettes, type Pellet, sans robinet intermédiaire, Schellbach
Burettes
Automatic burette, Pellet, class AS, DURAN® with blue graduation.
  • Lateral stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
  • Certified conformity, with dated batch identification
Certifications: Compliance with DIN EN ISO 385.