Imprimer
Condenseurs, Dimroth, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
242546101 242547106 242547209 242548205
SCOT242546101EA 122 EUR
SCOT242546101 SCOT242547106 201-3335 SCOT242548205
Condenseurs, Dimroth, DURAN®
Condenseurs
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. Water supplied through plastic connectors.
The Dimroth condenser comprises a coil condenser located within a tube. This condenser type has a larger heat exchange surface and thus a better cooling effect than the Liebig or Allihn condenser.