Imprimer
DURAN® Micro Filter Candles without Tube
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
258572107 258572201 258572304 258572407
SCOT258572107EA 606 EUR
SCOT258572107 SCOT258572201 SCOT258572304 SCOT258572407
DURAN® Micro Filter Candles without Tube
Filtres Tubes filtrants
These micro filter candles are made from clear DURAN® borosilicate glass 3.3.