Imprimer
Bonbonnes
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1781 1782 1783 1780
KART1781EA 564 EUR
KART1781 KART1782 216-1040 KART1780
Bonbonnes
Flacons Jerricans
HDPE, translucent with black screw cap.
  • With comfortable handle and secure seal cap following DIN recommendations
  • Conform to UN regulation on transportation of dangerous liquids
  • Suitable for foodstuff
Ideal for storing and transporting liquids.