Imprimer
Filter funnels, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
258520101 258520204 258520307 258520401 258520504 258521106 258521209 258521303 258521406 258521509 258522102 258522205 258522308 258522402 258522505 258523107 258523201 258523304 258523407 258523501 258524103 258524206 258524309 258524403 258524506 258526104 258526207 258526301 258526404 258526507
511-0227EA 63.5 EUR
511-0227 511-0226 511-0225 511-0224 511-0223 511-0222 511-0221 511-0220 511-0219 511-0099 SCOT258522102 SCOT258522205 SCOT258522308 SCOT258522402 SCOT258522505 SCOT258523107 SCOT258523201 SCOT258523304 SCOT258523407 SCOT258523501 SCOT258524103 SCOT258524206 SCOT258524309 SCOT258524403 SCOT258524506 SCOT258526104 SCOT258526207 SCOT258526301 SCOT258526404 SCOT258526507
Filter funnels, DURAN®
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.