Imprimer
Filter funnels, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
258540309 258540403
SCOT258540309EA 57.7 EUR
SCOT258540309 SCOT258540403
Filter funnels, DURAN®
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.