Imprimer
Bulb pipettes, class B, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
243370108 243370202 243370708 243370802 243371207 243371404 243371704 243372409
612-5234EA 11.6 EUR
612-5234 612-5235 612-5236 612-5237 612-5238 612-5239 612-5240 612-5241
Bulb pipettes, class B, DURAN®
Pipettes Pipettes jaugées
Soda-lime glass (AR-Glas®).
  • Marks and inscription in brown
  • Calibration is based on the poured out volume (Ex)
Complies to DIN ISO 648.
Colour code complies to DIN 12621.
*no bulb