Imprimer
Graduated pipettes, Class AS, total and partial delivery, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
243451109 243451709 243452302 243452902 243453401
612-5266EA 164 EUR
612-5266 612-5267 612-5268 612-5269 612-5270
Graduated pipettes, Class AS, total and partial delivery, DURAN®
Pipettes Pipettes graduées
Soda-lime glass (AR-Glas®).
  • Batch certificate and certificate of conformity
  • Printed Retrace Code (batch identification)
  • For cotton plugs
  • Marks and inscription in brown with ring graduations
  • Numbering: Zero at top
  • Graduated to tip (total delivery, type 3)
Complies to DIN ISO 835.
Colour code complies to DIN 12621.
Length: 360 mm for items with 25 ml or more 450 mm.