Imprimer
Graduated pipettes, Class B, partial delivery, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
243431604 243432309 243432909 243433408 243430608 243430308 243431107 243430102
612-5254EA 118 EUR
612-5254 612-5255 612-5256 612-5257 612-5252 612-5251 612-5253 612-5250
Graduated pipettes, Class B, partial delivery, DURAN®
Pipettes Graduated Pipettes
Soda-lime glass (AR-Glas®).
  • For cotton plugs
  • Marks and inscription in brown
  • Numbering: Zero at top
  • Partial delivery (type 1)
  • Calibration is based on the poured out volume (Ex)
Complies to DIN ISO 835.
Colour code complies to DIN 12621.
Certifications: ISO 835.