Imprimer
Burettes, robinet droit, classe B, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
243283605 243283905 243282703 243283305
612-5224EA 250 EUR
612-5224 612-5225 612-5222 612-5223
Burettes, robinet droit, classe B, DURAN®
Burettes
DURAN®, borosilicate glass 3.3, clear.
  • Straight stopcock
  • With glass key
  • Marks and inscription in blue
  • Calibration is based on the poured out volume ('Ex')
Class B limit is roughly one and a half times wider than for class AS. The tolerances are thus more strict than specified by DIN. Typical application: titrations.
Certifications: Complies to the DIN ISO 385.