Imprimer
Burettes PMP, robinet droit, clé FEP
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1997 1998 1999
KART1997EA 198 EUR
KART1997 612-2813 KART1999
Burettes PMP, robinet droit, clé FEP
Burettes
High precison plastic burettes particularly useful for routine work to eliminate the danger of glass breakages, provides good resistance to chemicals.
  • Combination of different plastic materials (PMP, FEP, PVC)
  • The stopcock assembly is moulded in one piece, with FEP key
  • Requiring no lubricant and providing a leak-proof seal
Certifications: Calibrated according to DIN 12.700 standards - Class B.