Imprimer
Bains
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20004382 20004383
IKAA20004382EA 766 EUR
IKAA20004382 IKAA20004383
Bains
Bains Bains-marie
Attention: Note: The product is a rectangular plastic bath without a thermostat, the thermostatic controller must be ordered separately.