Imprimer
Tubes à échantillons RMN, 5 mm, avec classes de précision, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
231700314 231700117 231700211
212-1682EA 138 EUR
212-1682 212-1680 SCOT231700211
Tubes à échantillons RMN, 5 mm, avec classes de précision, DURAN®
Tubes Tubes RMN
DURAN®, borosilicate glass 3.3, clear.
  • Storage and transport of measurement solutions
  • Designed for use in low to most mid field NMR spectrometers
  • Close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution
DURAN® NMR tubes are available, according to requirement, in three accuracy classes. The correct tube can be selected depending on resonant frequency. These tubes are noteworthy for their close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution. Consequently, quick and accurate test results are achievable.
With printed retrace code (batch identification).