Imprimer
Distilling receivers, Bredt, straight
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05285001 05285004 05285005
LENZ05285001EA 287 EUR
LENZ05285001 LENZ05285004 LENZ05285005
Distilling receivers, Bredt, straight
Composants de distillation Receveurs de distillation
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With four flasks NS 14/23, with straight adapter, vacuum connection: GL 18.