Imprimer
Low TOC cleaning validation swabs, Alpha®
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TX714K TX761K
115-1894EA 1680 EUR
115-1894 115-1898
Low TOC cleaning validation swabs, Alpha®
Écouvillons Écouvillons de nettoyage
Low TOC swabs are made from 100% polyester knit materials, which are further cleaned for use in Total Organic Carbon (TOC) analysis.
Suitable for surface sampling in validation protocols and precision cleaning for hard-to-reach areas and small spaces.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904.