Imprimer
Flacons de diagnostic, col étroit, ronds, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
362004-9125 362004-9050 362004-9025
NALG362004-9125EA 0 EUR
NALG362004-9125 NALG362004-9050 NALG362004-9025
Flacons de diagnostic, col étroit, ronds, Nalgene®
Flacons Flacons en plastique
Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ opaque amber HDPE diagnostic bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
  • Durable, non-sterile and leakproof
  • Non-pyrogenic and non-cytotoxic
  • Uniform walls are resistant to splitting or puncturing
Bottles provide light resistance and used in test kits and many small-volume applications.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Informations de commande: Product certificate providing cytotoxicity, USP Class VI, FDA Compliance - 21 CFR and other regulatory compliance information is provided based upon request.
Cdt: Bottles are bulk packed without closures for 2000 per case. Coloured closures should be order seperately.