Imprimer
Flacons de diagnostic à col étroit, ronds, avec bouchon à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
322002-9025 322002-9050 322002-9125
NALG322002-9025EA 1690 EUR
NALG322002-9025 NALG322002-9050 NALG322002-9125
Flacons de diagnostic à col étroit, ronds, avec bouchon à vis, Nalgene®
Flacons Flacons en plastique
Perform many small-volume applications with Thermo Scientific™ Nalgene™ HDPE diagnostic bottles with closure. These bottles are ideal for use in test kits, sample storage and shipping of small-volume, valuable reagents. They come packed in rigid trays for easy handling.
  • Leakproof and nonsterile
  • Natural polypropylene (PP) closure for excellent chemical resistance
  • Durable, uniform walls that are resistant to splitting or puncturing
Certifications: Compliance with ISO 13485:2003 quality requirements and 21CFR177.1520 for food and beverage use, USP Class VI
Cdt: Bottles are packed in shrink-wrapped trays and the closures are packed in a separate polybag within the same carton.