Imprimer
Flacons de diagnostic à col étroit, ronds, avec bouchon à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
322004-9025 322004-9050 322004-9125
NALG322004-9025EA 0 EUR
NALG322004-9025 NALG322004-9050 NALG322004-9125
Flacons de diagnostic à col étroit, ronds, avec bouchon à vis, Nalgene®
Flacons Flacons en plastique
Narrow mouth Boston round diagnostic bottle is made of opaque amber high-density polyethylene (HDPE) and comes with an opaque amber polypropylene closure.
  • Good chemical resistance
  • Good temperature resistance
Ideal for packaging light sensitive reagents; leakproof bottle has durable, uniform walls that resist splitting or puncturing.
Certifications: Manufactured in compliance with ISO 13485 quality requirements; meets USP requirements for light resistant containers.
Cdt: Packed in a rigid tray .