Imprimer
Tubes à essais
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1400 1405
KART1400EA 70 EUR
KART1400 KART1405
Tubes à essais
Tubes Tubes à essais
PTFE, white.
  • Excellent chemical resistance
  • Very good mechanical resistance
  • Very good temperature resistance
  • Suitable for foodstuff
Will withstand temperatures continuously up to 250 °C and for short intervals up to 300 °C.