Imprimer
Flacons de conditionnement, col étroit, ronds, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
362085-0001 362085-0004
NALG362085-0001EA 564 EUR
NALG362085-0001 NALG362085-0004
Flacons de conditionnement, col étroit, ronds, Nalgene®
Flacons Flacons en plastique
HDPE, opaque amber.
  • Excellent impact and chemical resistance
  • For light sensitive materials
  • Good temperature resistance
Supplied without a closure to assist in feeding into filling lines.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.