Imprimer
Flacons, col étroit, type Boston, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
362008-0008 362008-0001 362008-0002 362008-0016 362008-0032
NALG362008-0008EA 350 EUR
NALG362008-0008 NALG362008-0001 NALG362008-0002 NALG362008-0016 NALG362008-0032
Flacons, col étroit, type Boston, Nalgene®
Flacons Flacons en plastique
Bottles, narrow neck, made of HDPE, opaque white, without closure, round shape, non sterile. Specifically designed for demanding lab use, these reliable, durable bottles meet the specifications of light-resistant containers per USP requirements, latest edition.
  • Durable, uniform walls that are resistant to splitting or puncturing
  • Rigid and translucent, with excellent chemical resistance
  • Good for freezer use to −100 °C
  • Leakproof when used with nalgene closures
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. BottlesEn savoir plus