Imprimer
Bottles, wide neck, with standard ground joints, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
211847306 211848602 211849101 211845408 211844403 211846301 211842402 211843604
SCOT211847306EA 352 EUR
SCOT211847306 SCOT211848602 SCOT211849101 SCOT211845408 SCOT211844403 SCOT211846301 SCOT211842402 SCOT211843604
Bottles, wide neck, with standard ground joints, DURAN®
Flacons Flacons en verre
DURAN®, borosilicate glass 3.3, clear or amber, with standard ground joint.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Without stopper. Marked with volume and equipped with a white labelling field. Amber to protect light-sensitive materials.
With printed retrace code (batch identification).
Certifications: ISO 4796-2