Imprimer
Potassium dihydrogen citrate hydrate ≥99% (base substance sèche)
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Potassium citrate monobasic hydrate, Citric acid monopotassium salt hydrate, mono-Potassium citrate hydrate
44415.36 44415.A3
44415.36EA 110 EUR
44415.36 44415.A3
Potassium dihydrogen citrate hydrate ≥99% (base substance sèche)
Potassium dihydrogen citrate hydrate
Potassium dihydrogen citrate hydrate is used as a buffering agent and a chelating agent. It is used as an alkalizing agent in the renal function. It is found that it increases the bone density, thereby used in research experiments related to the treatment of osteoporosis.
Purity: 99% CAS No.: 866-83-1 Molecular Formula: HOOCCH2C(OH)(COOH)CH2COOK.xH2O Formula Weight: 230.22(anhy) Physical Form: Powder/ chunks Sensitivity: Hygroscopic Beilstein Reference: 4772973 Merck Reference: 14,7624 MDL No.: MFCD00036472
Formule: C₆H₇KO₇
Poids moléculaire: 230.22 g/mol (anhy)
Température de stockage: Température ambiante
Numéro MDL: MFCD00036472
Numéro CAS: 866-83-1
EINECS: 212-753-4
Indice Merck: 14,7624