Imprimer
Flacons, col large, PEHD, ronds, avec bouchon à vis, qualité laboratoire, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312104-0008 312104-0004 312104-0002 312104-0001 312104-0016 312104-0032
215-2768EA 564 EUR
215-2768 215-2767 215-2766 215-2574 215-2769 215-2770
Flacons, col large, PEHD, ronds, avec bouchon à vis, qualité laboratoire, Nalgene®
Flacons Flacons en plastique
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof closure
Can be used anywhere in the laboratory and for sample collection. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Cdt: Bottles and closures are packed in separate polybags within the same carton.