Imprimer
Flacons, col étroit, fluorés, avec bouchon à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312097-0004 312097-0008 312097-0016 312097-0032
NALG312097-0004EA 1250 EUR
NALG312097-0004 NALG312097-0008 NALG312097-0016 NALG312097-0032
Flacons, col étroit, fluorés, avec bouchon à vis, Nalgene®
Flacons Flacons en plastique
HDPE, fluorinated with PP, fluorinated screw cap.
  • Fluorinated bottles and carboys can be used for most acids, alkalis and aggressive organic solvents
  • Round shape, non sterile
  • Heavy-duty walls are safe, durable and resistant to splitting and punctures
  • Fluorocarbon surfaces inside and out
  • Leakproof
Fluorinating these HDPE bottles and carboys provides a cost effective alternative to costly fluoropolymer containers.
Manufactured from Nalgene™ for resistance to permeation, paneling, odor emission, flavor and fragrance loss. Provide superior recepticals for solvents, oils, chemicals and agricultural products.
Certifications: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP class VI.