Imprimer
Bellow connectors
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1047 1048 1049
KART1047EA 229 EUR
KART1047 KART1048 KART1049
Bellow connectors
Adaptateurs Raccords à rodage conique
Bellow connector, PTFE, for use as a flexible or elastic connector to prevent pressure and stresses, and to dampen vibrations in standard ground joints.
  • Inert
  • Temperature resistant up to 280 °C
  • Very good chemical resistance
With ground joints on cone and socket. Sturdy construction with trapezoidal sealing rings, which give medium to high vacuum performance.