Imprimer
Sleeves for conical joints
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1040 1035 1036 1037 1038 1039 1041 1042 1043
KART1040EA 58.6 EUR
KART1040 KART1035 KART1036 KART1037 KART1038 KART1039 KART1041 KART1042 KART1043
Sleeves for conical joints
Manchons et raccords pour joints NS Manchettes pour rodage de raccordement
PTFE.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight (to 0,1 mbar)
  • Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.